missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPCD Polyclonal Antibody
GREENER_CHOICE

Product Code. 15745631 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15745631 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15745631 Supplier Invitrogen™ Supplier No. PA557738

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: YYKKFSIPDL DRHQLPLDDA LLSFAHANCT LIISYQKPKE VVVAESELQK ELKKVKTAHS NDGDCK Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 91%.

May play a role in the formation or function of ciliated cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPCD
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DPCD
Gene Accession No. Q9BVM2
Gene Alias 5330431N19Rik; deleted in a mouse model of primary ciliary dyskinesia; deleted in primary ciliary dyskinesia; deleted in primary ciliary dyskinesia homolog (mouse); DPCD; Ndac; neighbor of Dac; protein DPCD; RGD1307648; RP11-529I10.4; RP11-529I104
Gene Symbols DPCD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human DPCD. Recombinant protein control fragment (Product #RP-102728).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25911
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.