Learn More
Lovibond™ Replacement Reagent for CHECKIT™ Comparator
Description
Lovibond™ Replacement Reagent for CHECKIT™ Comparator is manufactured under tightly controlled conditions to maintain the highest quality standards.
- Highest quality production standards
- Consistent and reliable reagents are provided in liquid and tablet formats
- Maximum accuracy
Specifications
Specifications
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | Colorimeter and spectrophotometer |
| Quantity | 250 Pack |
| Type | DPD Glycine Test Tablets (for Cl separation) |