missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPF2 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76512JF646
This item is not returnable.
View return policy
Description
DPF2 Polyclonal antibody specifically detects DPF2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DPF2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Janelia Fluor 646 | |
| Apoptosis response zinc finger protein, D4, zinc and double PHD fingers family 2BAF45D, MGC10180, Protein requiem, REQBRG1-associated factor 45D, requiem, apoptosis response zinc finger, ubi-d4, UBID4requiem, apoptosis response zinc finger gene, zinc finger protein ubi-d4 | |
| Antibody was raised against an 18 amino acid synthetic peptide near the carboxy terminus of human Requiem. The immunogen is located within the last 50 amino acids of REQUIEM. Amino Acid Squence: LDLLKEKASIYQNQNSS | |
| 0.1 mL | |
| Apoptosis | |
| 5977 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction