missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPIL4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35619-20ul
This item is not returnable.
View return policy
Description
PPIL4 Polyclonal antibody specifically detects PPIL4 in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/Immunofluorescence
Specifications
| PPIL4 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 | |
| Cyclophilin-like protein PPIL4, cyclophilin-type peptidyl-prolyl cis-trans isomerase, EC 5.2.1.8, HDCME13P, peptidyl-prolyl cis-trans isomerase-like 4, peptidylprolyl isomerase (cyclophilin)-like 4, PPIase, Rotamase PPIL4, serologically defined breast cancer antigen NY-BR-18 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 301-492 of human PPIL4 (NP_624311.1).,, Sequence:, MDNVLIDDRRIHVDFSQSVAKVKWKGKGGKYTKSDFKEYEKEQDKPPNLVLKDKVKPKQDTKYDLILDEQAEDSKSSHSHTSKKHKKKTHHCSEEKEDEDYMPIKNTNQDIYREMGFGHYEEEESCWEKQKSEKRDRTQNRSRSRSRERDGHYSNSHKSKYQTDLYERERSKKRDRSRSPKKSKDKEKSKYR | |
| 20 μL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 85313 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction