missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPH2 Antibody (OTI4E7), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70585IR
This item is not returnable.
View return policy
Description
DPH2 Monoclonal specifically detects DPH2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.
Specifications
| DPH2 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DPH2 | |
| Full length recombinant protein of human DPH2 (NP_001375) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG1 |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| OTI4E7 | |
| Western Blot, Flow Cytometry, Immunohistochemistry | |
| diphthamide biosynthesis protein 2, Diphthamide biosynthesis protein 2 homolog-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2, diptheria toxin resistance protein required for diphthamide biosynthesis-like 2(S. cerevisiae), DPH2 homolog, DPH2 homolog (S. cerevisiae), DPH2L2diphthamide biosynthesis-like protein 2, DPH2-like 2, DPH2-like 2 (S. cerevisiae), HsDph2 | |
| Mouse | |
| Affinity Purified | |
| Cell Cycle and Replication | |
| 1802.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction