missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DPH2 Monoclonal Antibody (OTI4E7)
GREENER_CHOICE

Product Code. 15915594
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15915594 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15915594 Supplier Invitrogen™ Supplier No. MA525994

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DPH2 Monoclonal Antibody for Western Blot, IHC (P), Flow

This gene is one of two human genes similar to the yeast gene dph2. The yeast gene was identified by its ability to complement a diphthamide mutant strain, and thus probably functions in diphthamide biosynthesis. Diphthamide is a post-translationally modified histidine residue present in elongation factor 2 (EF2) that is the target of diphtheria toxin ADP-ribosylation. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen DPH2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI4E7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene DPH2
Gene Accession No. Q568Y2, Q9BQC3, Q9CR25
Gene Alias 2-(3-amino-3-carboxypropyl)histidine synthase subunit 2; 9130020C19Rik; AI467389; diphthamide biosynthesis 2; diphthamide biosynthesis protein 2; diphthamide biosynthesis protein 2 homolog-like 2; diphthamide biosynthesis-like protein 2; Diphtheria toxin resistance protein 2; diptheria toxin resistance protein required for diphthamide biosynthesis (Saccharomyces)-like 1; diptheria toxin resistance protein required for diphthamide biosynthesis (Saccharomyces)-like 2; diptheria toxin resistance protein required for diphthamide biosynthesis-like 2; DPH2; DPH2 homolog; DPH2 homolog (S. cerevisiae); DPH2L2; DPH2-like 2; EGK_00671; MmDph2; S-adenosyl-L-methionine:L-histidine 3-amino-3-carboxypropyltransferase 2; S-adenosyl-L-methionine:L-histidine 3-amino-3-carboxypropyltransferase DPH2
Gene Symbols DPH2
Host Species Mouse
Immunogen Full length human recombinant protein of DPH2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1802, 298452, 475387, 67728
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.