missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPH4 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99139CL1
This item is not returnable.
View return policy
Description
DPH4 Polyclonal antibody specifically detects DPH4 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| DPH4 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| CSL-type zinc finger-containing protein 3, DnaJ (Hsp40) homolog, subfamily C, member 24, dnaJ homolog subfamily C member 24, DPH4 homolog, DPH4, JJJ3 homolog, DPH4, JJJ3 homolog (S. cerevisiae), DPH41700030A21Rik, JJJ3, S. cerevisiae), ZCSL3, zinc finger, CSL domain containing 3, zinc finger, CSL-type containing 3 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DPH4. | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| 120526 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction