missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPH4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | DPH4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 5-10 μL/mg of lysate |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18697042
|
Novus Biologicals
NBP2-97599-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18615944
|
Novus Biologicals
NBP2-97599-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DPH4 Polyclonal antibody specifically detects DPH4 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| DPH4 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.0) | |
| 120526 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:2000, Immunoprecipitation 5-10 μL/mg of lysate | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CSL-type zinc finger-containing protein 3, DnaJ (Hsp40) homolog, subfamily C, member 24, dnaJ homolog subfamily C member 24, DPH4 homolog, DPH4, JJJ3 homolog, DPH4, JJJ3 homolog (S. cerevisiae), DPH41700030A21Rik, JJJ3, S. cerevisiae), ZCSL3, zinc finger, CSL domain containing 3, zinc finger, CSL-type containing 3 | |
| Produced in rabbits immunized with E. coli-derived Human DPH4 fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title