missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98802-100ul
This item is not returnable.
View return policy
Description
DPM1 Polyclonal antibody specifically detects DPM1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DPM1 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| CDGIEEC 2.4.1.83, dolichol monophosphate mannose synthase, dolichol-phosphate mannosyltransferase, Dolichyl-phosphate beta-D-mannosyltransferase, dolichyl-phosphate mannosyltransferase polypeptide 1, catalytic subunit, DPM synthase, Mannose-P-dolichol synthase, MPD synthase, MPDSDolichol-phosphate mannose synthase | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human DPM1. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 8813 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction