missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPM3 Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-26105
This item is not returnable.
View return policy
Description
DPM3 Polyclonal specifically detects DPM3 in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| DPM3 | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| DPM3 | |
| Peptide with sequence HDCEDAARELQSQ corresponding to internal region according to NP_061846.2, NP_714963.1. | |
| 0.1 mg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_061846.2 and NP_714963.1). | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 1 - 3 ug/ml, Peptide ELISA Detection limit 1:1000 | |
| CDG1O, Dolichol-phosphate mannose synthase subunit 3, dolichol-phosphate mannosyltransferase subunit 3, Dolichyl-phosphate beta-D-mannosyltransferase subunit 3, dolichyl-phosphate mannosyltransferase polypeptide 3, DPM synthase complex subunit 3, Mannose-P-dolichol synthase subunit 3, MGC125904, MGC125905, MGC34275, MPD synthase subunit 3, prostin 1, prostin-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 54344 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction