missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP10 Antibody (004), Alexa Fluor™ 488 , Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89978AF488
This item is not returnable.
View return policy
Description
DPP10 Monoclonal antibody specifically detects DPP10 in Human samples. It is validated for ELISA
Specifications
| DPP10 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| dipeptidyl peptidase 10, Dipeptidyl peptidase IV-related protein 3, dipeptidyl peptidase like protein 2, Dipeptidyl peptidase X, Dipeptidyl peptidase-like protein 2, dipeptidyl-peptidase 10, dipeptidyl-peptidase 10 (non-functional), DPL2, DPP X, DPPY, DPRP-3, DPRP3dipeptidylpeptidase 10, inactive dipeptidyl peptidase 10, KIAA1492 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human DPP10 (Q8N608-1; Leu 56-Glu 796). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 004 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 57628 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction