missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP3 Antibody (986613), Alexa Fluor™ 594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals MAB8087AF594
This item is not returnable.
View return policy
Description
DPP3 Monoclonal specifically detects DPP3 in Human samples. It is validated for Western Blot.
Specifications
| DPP3 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DPP3 | |
| E. coli-derived recombinant human DPP3 protein (amino acids 2-737) | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| Human | |
| IgG2a |
| Western Blot | |
| 986613 | |
| Western Blot | |
| Dipeptidyl aminopeptidase III, Dipeptidyl arylamidase III, dipeptidyl peptidase 3, Dipeptidyl peptidase III, dipeptidylpeptidase 3, dipeptidyl-peptidase 3, dipeptidylpeptidase III, DPP III, DPPIII, EC 3.4.14.4, FLJ11387, FLJ22331 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction