missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPP3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5122028
This item is not returnable.
View return policy
Description
DPP3 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a protein that is a member of the M49 family of metallopeptidases. This cytoplasmic protein binds a single zinc ion with its zinc-binding motif (HELLGH) and has post-proline dipeptidyl aminopeptidase activity, cleaving Xaa-Pro dipeptides from the N-termini of proteins. Increased activity of this protein is associated with endometrial and ovarian cancers. Alternate transcriptional splice variants have been characterized.
Specifications
| DPP3 | |
| Polyclonal | |
| Unconjugated | |
| DPP3 | |
| 4930533O14Rik; C86324; Dipeptidyl aminopeptidase III; dipeptidyl arylamidase III; Dipeptidyl peptidase 3; dipeptidyl peptidase III; dipeptidylpeptidase 3; dipeptidyl-peptidase 3; dipeptidylpeptidase III; dipeptidyl-peptidase III; DPP III; Dpp3; DPPIII; Enkephalinase B; hCG_2016942 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100519350, 10072, 114591 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| O55096, Q9NY33 | |
| DPP3 | |
| Recombinant protein Dipeptidyl Peptidase 3. The antigen corresponds to amino acid range 2-261 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction