missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPP6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA533473
This item is not returnable.
View return policy
Description
Percent identity with other species by BLAST analysis: Human, Chimpanzee, Gorilla, Gibbon, Monkey, Marmoset, Mouse, Rat, Elephant, Panda, Bovine, Horse, Rabbit, Xenopus (100%).
DPP6 encodes a single-pass type II membrane protein that is a member of the S9B family in clan SC of the serine proteases. This protein has no detectable protease activity, most likely due to the absence of the conserved serine residue normally present in the catalytic domain of serine proteases. However, it does bind specific voltage-gated potassium channels and alters their expression and biophysical properties. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications
| DPP6 | |
| Polyclonal | |
| Unconjugated | |
| DPP6 | |
| B930011P16Rik; D5Buc3; D5Buc4; D5Buc5; dipeptidyl aminopeptidase IV-related protein; dipeptidyl aminopeptidase-like protein 6; Dipeptidyl aminopeptidase-related protein; Dipeptidyl peptidase 6; Dipeptidyl peptidase IV-like protein; dipeptidyl peptidase IV-related protein; dipeptidyl peptidase like 6; dipeptidyl peptidase VI; dipeptidylpeptidase 6; dipeptidyl-peptidase 6; dipeptidylpeptidase VI; DPL1; DPP VI; Dpp6; Dpp-6; DPPX; Gm1377; In(5)6H p; In(5)6H-p; inversion, Chr 5, Harwell 6, proximal; MRD33; Peplb; Rw; VF2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100063666, 100344997, 13483, 1804, 281123, 29272 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| P42658, P42659, P46101, Q9Z218 | |
| DPP6 | |
| Synthetic 18 amino acid peptide from extracellular domain of human DPP6. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Monkey, Bovine, Horse, Frog | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction