missing translation for 'onlineSavingsMsg'
Learn More

DPP9 Monoclonal Antibody (OTI2F10), TrueMAB™, OriGene

Product Code. 17500372
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17500372 30 μL 30µL
17580362 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17500372 Supplier Origene Technologies Supplier No. TA503937S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

DPP9 Monoclonal antibody specifically detects DPP9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen DPP9
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI2F10
Concentration 0.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene PDSS2
Gene Accession No. Q86YH6
Gene Alias 6430584G11Rik; A330078I11; Dipeptidyl peptidase 9; dipeptidyl peptidase IV-related protein 2; dipeptidyl peptidase IV-related protein-2; dipeptidyl peptidase IX; Dipeptidyl peptidase-like protein 9; dipeptidylpeptidase 9; dipeptidyl-peptidase 9; DKFZp762F117; DP9; DPLP9; DPP IX; DPP9; DPRP2; DPRP-2; FLJ16073
Host Species Mouse
Immunogen Full length human recombinant protein of human PDSS2 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57107
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.