missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPP9 Monoclonal Antibody (OTI6A12), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504039S
This item is not returnable.
View return policy
Description
DPP9 Monoclonal antibody specifically detects DPP9 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| DPP9 | |
| Monoclonal | |
| 0.86 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P16406, Q07075 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 13809, 2028 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI6A12 | |
| Unconjugated | |
| Enpep | |
| 6430584G11Rik; A330078I11; Dipeptidyl peptidase 9; dipeptidyl peptidase IV-related protein 2; dipeptidyl peptidase IV-related protein-2; dipeptidyl peptidase IX; Dipeptidyl peptidase-like protein 9; dipeptidylpeptidase 9; dipeptidyl-peptidase 9; DKFZp762F117; DP9; DPLP9; DPP IX; DPP9; DPRP2; DPRP-2; FLJ16073 | |
| Full length human recombinant protein of human ENPEP produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human, Mouse | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction