missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPA2 Antibody (OTI1G10), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72426R
This item is not returnable.
View return policy
Description
DPPA2 Monoclonal specifically detects DPPA2 in Human, Rat, Canine samples. It is validated for Western Blot.
Specifications
| DPPA2 | |
| Monoclonal | |
| DyLight 550 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DPPA2 | |
| Recombinant protein fragment corresponding to amino acids 1-175 of human DPPA2 (NP_620170) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Rat, Canine | |
| IgG1 |
| Western Blot | |
| OTI1G10 | |
| Western Blot | |
| CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein | |
| Mouse | |
| Affinity Purified | |
| Developmental Biology, Stem Cells | |
| 151871.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction