missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPA2 Antibody (Z31P3C4*F11), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50230UV
This item is not returnable.
View return policy
Description
DPPA2 Monoclonal specifically detects DPPA2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| DPPA2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| DPPA2 | |
| Peptide Sequence -KMMKRLMTVE | |
| 0.1 mL | |
| Developmental Biology, Stem Cells | |
| 151871 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry | |
| Z31P3C4*F11 | |
| Western Blot, ELISA, Immunohistochemistry | |
| CT100, developmental pluripotency associated 2, developmental pluripotency-associated protein 2, PESCRG1cancer/testis antigen 100, Pluripotent embryonic stem cell-related gene 1 protein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction