missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPPA2 Monoclonal Antibody (OTI1G10)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526429
This item is not returnable.
View return policy
Description
DPPA2 Monoclonal Antibody for Western Blot
The DPPA2 (Developmental Pluripotency Associated 2) gene is prominently expressed in embryonic stem cells and is critical for maintaining their pluripotency and self-renewal capabilities. Located on chromosome 3, DPPA2 belongs to a family of genes that play significant roles in early embryonic development and epigenetic remodeling. The expression of DPPA2 is tightly regulated through mechanisms such as DNA methylation, and it is involved in regulatory pathways that control the expression of core pluripotency factors. Beyond its role in stem cell biology, DPPA2 expression is also noted in various cancers, where it can become reactivated, suggesting its involvement in oncogenic processes. In the context of cancer, DPPA2, along with DPPA4, has been studied for its potential to facilitate cellular reprogramming and contribute to tumorigenesis. Given its expression pattern, DPPA2 serves as both a marker of pluripotent potential in stem cells and a potential target for cancer diagnostics and therapy due to its reactivation in malignant tissues.
Specifications
| DPPA2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7Z7J5 | |
| DPPA2, LOC683282 | |
| Human recombinant protein fragment corresponding to amino acids 1-175 of DPPA2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI1G10 | |
| Unconjugated | |
| DPPA2 | |
| 2410088E07Rik; C80932; cancer/testis antigen 100; CT100; D19Mgi18; developmental pluripotency associated 2; developmental pluripotency-associated 2; developmental pluripotency-associated protein 2; DPPA2; ECAT15-2; ECSA; embryonic stem cell (ESC) associated transcript 15-2; embryonic stem cell-associated transcript 15-2 protein; LOC683282; PESCRG1; Phsecrg1; Pluripotent embryonic stem cell-related gene 1 protein; similar to developmental pluripotency-associated 2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 151871, 607166, 683282 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction