missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPA4 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99578-100ul
This item is not returnable.
View return policy
Description
DPPA4 Polyclonal antibody specifically detects DPPA4 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| DPPA4 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:500-1:2000 | |
| developmental pluripotency associated 4, developmental pluripotency-associated protein 4, FLJ10713,2410091M23Rik | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human DPPA4 | |
| 100 μg | |
| Stem Cell Markers | |
| 55211 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.0) | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction