Learn More
DPPIV/CD26 Antibody (6G7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26613-50ul
 View more versions of this product
Product Code. 30043493
  • € 343.00 / 50µL
Description
DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Human samples. It is validated for ELISA, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DPPIV/CD26
Monoclonal
Unconjugated
PBS, pH 7.4, 150mM NaCl and 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
ELISA, Immunohistochemistry
6G7
ELISA, Immunohistochemistry 1:50-1:200
ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5
A synthesized peptide derived from Human DPPIV/CD26 [UniProt P27487]
50 μL
Adaptive Immunity, Cellular Markers, GPCR, Immunology
1803
Store at -20 to -70C. Avoid freeze-thaw cycles.