missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DPPIV/CD26 Antibody (DPP4/910), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47912AF405
This item is not returnable.
View return policy
Description
DPPIV/CD26 Monoclonal antibody specifically detects DPPIV/CD26 in Human, Rat, Porcine (Negative), Sheep (Negative) samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen), Flow (Cell Surface), Immunofluorescence, CyTOF
Specifications
| DPPIV/CD26 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| ADABP, ADCP-2, ADCP2DPP IV, Adenosine deaminase complexing protein 2TP103, CD26 antigen, CD26T-cell activation antigen CD26, dipeptidyl peptidase 4, Dipeptidyl peptidase IV, dipeptidylpeptidase 4, dipeptidyl-peptidase 4, dipeptidylpeptidase IV (CD26, adenosine deaminase complexing protein 2), DPPIV, EC 3.4.14.5 | |
| Recombinant human DPPIV/CD26 protein (Uniprot: P27487) | |
| 0.1 mL | |
| Adaptive Immunity, Cellular Markers, Immunology | |
| 1803 | |
| Store at 4°C in the dark. | |
| IgG2b κ |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Flow Cytometry (Cell Surface) | |
| DPP4/910 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Rat, Porcine (Negative), Sheep (Negative) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction