missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ DPY19L4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA528515
This item is not returnable.
View return policy
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (94%), Rat (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Probable C-mannosyltransferase that mediates C-mannosylation of tryptophan residues on target proteins. [UniProt].
Specifications
| DPY19L4 | |
| Polyclonal | |
| Unconjugated | |
| DPY19L4 | |
| dpy-19 like 4 (C. elegans); DPY19L4; dpy-19-like 4; dpy-19-like 4 (C. elegans); dpy-19-like protein 4; Gm1023; Narg3; NMDA receptor-regulated gene 3; probable C-mannosyltransferase DPY19L4; Protein dpy-19 homolog 4; RGD1305828 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 286148 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q7Z388 | |
| DPY19L4 | |
| Recombinant protein encompassing a sequence within the C-terminus region of human DPY19L4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction