Learn More
DPY30 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32201DL594
This item is not returnable.
View return policy
Description
DPY30 Polyclonal antibody specifically detects DPY30 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)