Learn More
Invitrogen™ DPYSL3 Recombinant Rabbit Monoclonal Antibody (24GB3720)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557016
 View more versions of this product
Product Code. 30276615
  • € 151.00 / 20µL
Description
DPYSL3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
DPYSL3
Recombinant Monoclonal
Unconjugated
DPYSL3
Collapsin response mediator protein 4; collapsin response mediator protein 4 long; Crmp4; CRMP-4; dihydropyrimidinase like 3; dihydropyrimidinase-like 3; dihydropyrimidinase-related protein 3; Dpysl3; DRP3; DRP-3; LCRMP; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; TUC4; tuc-4; TUC-4b; ULIP; Ulip1; ULIP-1; Unc-33-like phosphoprotein 1
Rabbit
Affinity chromatography
RUO
1809, 22240, 25418
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3720
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q14195, Q62188, Q62952
DPYSL3
A synthesized peptide derived from human CRMP4 (aa 450-500).
20 μL
Primary
Human, Mouse, Rat