Learn More
Invitrogen™ DPYSL3 Recombinant Rabbit Monoclonal Antibody (24GB3720)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557016
This item is not returnable.
View return policy
Description
DPYSL3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
DPYSL3 is necessary for signaling by class 3 semaphorins and subsequent remodeling of the cytoskeleton. DPYSL3 plays a role in axon guidance, neuronal growth cone collapse and cell migration.
Specifications
| DPYSL3 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DPYSL3 | |
| Collapsin response mediator protein 4; collapsin response mediator protein 4 long; Crmp4; CRMP-4; dihydropyrimidinase like 3; dihydropyrimidinase-like 3; dihydropyrimidinase-related protein 3; Dpysl3; DRP3; DRP-3; LCRMP; testicular secretory protein Li 7; TOAD-64/Ulip/CRMP; TOAD-64/Ulip/CRMP protein 4b; TUC4; tuc-4; TUC-4b; ULIP; Ulip1; ULIP-1; Unc-33-like phosphoprotein 1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 1809, 22240, 25418 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB3720 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q14195, Q62188, Q62952 | |
| DPYSL3 | |
| A synthesized peptide derived from human CRMP4 (aa 450-500). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |