Learn More
Dräger™ Short-Term Detector Tubes - Hydrogen Sulfide
Product Code. 10378421
Click to view available options
Components Measured:
Hydrogen Sulfide 0.5/a
Hydrogen Sulfide 1/d
Hydrogen Sulfide 100/a
Hydrogen Sulfide 5/b
Measurement Range:
0.5 to 15ppm / 5 to 150ppm
1 to 200ppm
100 to 2000ppm
5 to 60ppm
Unit Size:
Pack of 10
Product Code. 10378421
Brand: Dräger™ CH29801
Specifications
Components Measured Hydrogen Sulfide 5/b
Humidity Range Less than40mg/L
Measurement Range 5 to 60ppm
No. of Strokes 10
Quantity 10
Response Time 4 min.
Temperature Range (English) 32°F to 140°F
Temperature Range (Metric) 0°C to 60°C
compliance-icons
Product Identifier
  • X10 GAS DETECTOR TUBES DRAEGER CH29801 HYDROGEN
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen sulphide