Learn More
Dräger™ Short-Term Detector Tubes - Monostyrene
Brand: Dräger™ CH27601
Additional Details : Weight : 0.07560kg
Description
1Extension of measuring range listed is possible for some detector tubes. Call Fisher Scientific™ at 1-800-772-6733 for more information.
2Safety Equipment Institute (SEI) certified.
Specifications
Monostyrene 50/a | |
50 to 400ppm | |
10 | |
50°F to 104°F |
Less than15mg/L | |
2 to 11 | |
2 min. | |
10° to 40°C |