missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR1 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-03329UV
This item is not returnable.
View return policy
Description
DR1 Polyclonal antibody specifically detects DR1 in Human samples. It is validated for Western Blot
Specifications
| DR1 | |
| Polyclonal | |
| Western Blot | |
| Down-regulator of transcription 1, down-regulator of transcription 1, TBP-binding (negative cofactor 2), NC2, NC2-BETA, Negative cofactor 2-beta, protein Dr1, TATA-binding protein-associated phosphoprotein | |
| The immunogen recognized by this antibody maps to a region between residue 25 and 75 of human down-regulator of transcription 1 using the numbering given in entry NP_001929.1 (GeneID 1810). | |
| 0.1 mL | |
| Stem Cell Markers | |
| 1810 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction