Learn More
Invitrogen™ DR6 Recombinant Rabbit Monoclonal Antibody (001)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA531266
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human DR6/TNFRSF21.
DR6 is widely expressed in human tissues and cell lines. The ligand for DR6 has not been identified. Apoptosis is induced by certain cytokines including TNF and Fas ligand of the TNF family through their death domain containing receptors, TNF-R1 and Fas. Several novel death receptors including DR3, DR4, and DR5 were recently identified. A new death domain containing receptor in the TNFR family was cloned recently and termed DR6 for death receptor-6. Like TNF-R1, DR6 interacts with death domain containing adapter molecule TRADD. Overexpression of DR6 induces apoptosis and activates NF-kappa-B and JNK.
Specifications
| DR6 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| O75509 | |
| TNFRSF21 | |
| Recombinant Human DR6 protein (Met1-Leu350). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 001 | |
| Unconjugated | |
| TNFRSF21 | |
| AA959878; BM-018; CD358; Death receptor 6; Death receptor-6; DR6; MGC31965; R74815; sCD358; sDR6; soluble CD358; soluble DR6; TNF receptor superfamily member 21; TNFR-related death receptor 6; TNFR-related death receptor-6; Tnfrsf21; TR7; Tumor necrosis factor receptor superfamily member 21; tumor necrosis factor receptor superfamily, member 21; UNQ437/PRO868 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 27242 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.