missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DR6/TNFRSF21 Rabbit anti-Human, Clone: 1, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 367.00 - € 552.00
Specifications
| Antigen | DR6/TNFRSF21 |
|---|---|
| Clone | 1 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18218657
|
Novus Biologicals
NBP2-89313-50UL |
50 μg |
€ 367.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18224846
|
Novus Biologicals
NBP2-89313-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
DR6/TNFRSF21 Monoclonal specifically detects DR6/TNFRSF21 in Human samples. It is validated for ELISA.Specifications
| DR6/TNFRSF21 | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| BM-018, CD358 antigen, Death receptor 6, DR6TNFR-related death receptor 6, MGC31965, tumor necrosis factor receptor superfamily member 21, tumor necrosis factor receptor superfamily, member 21 | |
| Recombinant Human DR6/TNFRSF21 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Cell Biology | |
| 0.2 um filtered solution in PBS | |
| 27242 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title