missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAK1 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76896AF532
This item is not returnable.
View return policy
Description
DRAK1 Polyclonal antibody specifically detects DRAK1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| DRAK1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| Alexa Fluor 532 | |
| DAP kinase-related apoptosis-inducing protein kinase 1, DRAK1death-associated protein kinase-related 1, EC 2.7.11.1, serine/threonine kinase 17a, serine/threonine kinase 17a (apoptosis-inducing), serine/threonine-protein kinase 17A | |
| Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human DRAK1. The immunogen is located within the first 50 amino acids of DRAK1. Amino Acid Squence: EKPGSGGSSPGATSG | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Signal Transduction | |
| 9263 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction