missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAK1 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-42692C
This item is not returnable.
View return policy
Description
DRAK1 Polyclonal antibody specifically detects DRAK1 in Human samples. It is validated for Western Blot
Specifications
| DRAK1 | |
| Polyclonal | |
| Western Blot | |
| DAP kinase-related apoptosis-inducing protein kinase 1, DRAK1death-associated protein kinase-related 1, EC 2.7.11.1, serine/threonine kinase 17a, serine/threonine kinase 17a (apoptosis-inducing), serine/threonine-protein kinase 17A | |
| The immunogen for this product maps to a region between residue 314 and 364 of human DAP kinase-related apoptosis-inducing protein kinase 1 using the numbering given in entry NP_004751.2 (GeneID 9263). | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Signal Transduction | |
| 9263 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| DyLight 650 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction