missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRAM Antibody (M3-P4B4), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50379B
This item is not returnable.
View return policy
Description
DRAM Monoclonal specifically detects DRAM in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| DRAM | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| DRAM1 | |
| Peptide Sequence - EKDYVYHVVS (amino acids 192 - 201) | |
| 0.1 mL | |
| Cancer | |
| 55332 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| M3-P4B4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| Damage-regulated autophagy modulator, DNA damage-regulated autophagy modulator protein 1, DNA-damage regulated autophagy modulator 1, DRAMFLJ11259 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction