missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Draxin/C1orf187 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 221.00 - € 305.00
Specifications
| Antigen | Draxin/C1orf187 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
Draxin/C1orf187 Polyclonal antibody specifically detects Draxin/C1orf187 in Human samples. It is validated for Western Blot, ELISASpecifications
| Draxin/C1orf187 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| 0.2 um filtered solution in PBS | |
| 374946 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AGPA3119, C1orf187, chromosome 1 open reading frame 187, dorsal inhibitory axon guidance protein, Dorsal repulsive axon guidance protein, draxin, FLJ34999, MGC117222, neucrin, UNQ3119 | |
| Produced in rabbits immunized with purified, recombinant Human Draxin/C1orf187 (Accession#: NP_940947.3; Met1-Val349) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title