Learn More
DRD2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 55084-1-AP-20UL
 View more versions of this product
Product Code. 16896525
  • € 145.00 / 20µL
Description
The antibody is specific to DRD2.
The DRD2 Dopamine Receptor is involved in the neuromodulation of appetitive behaviors. DRD2 may be linked to alcoholism and other substance-use disorders. Mutations in DRD2 in mice lead to the suppression of reward behavior with morphine and to the reduction of spontaneous movements, a phenotype resembling Parkinsons disease. Three alternatively spliced isoforms have been identified. DRD2 has been reported to be expressed in various regions of the brain, as well as in aorta, artery, eye, and spinal cord. ESTs have been isolated from normal pituitary and in brain, colon, lung, pancreas, and skeletal muscle cancer libraries.
Specifications
Specifications
DRD2
Polyclonal
Unconjugated
DRD2
D(2) dopamine receptor, D2DR, D2R, Dopamine D2 receptor, dopamine receptor D2, DRD2
Rabbit
Antigen Affinity Chromatography
RUO
13489, 1813, 24318
-20°C
Liquid
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin)
0.42 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14416, P61168, P61169
DRD2
Peptide
20 μL
Primary
Mouse, Rat, Human