missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drebrin 1 Antibody (DBN-N-03), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22344AF750
This item is not returnable.
View return policy
Description
Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| Drebrin 1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| DBN1 | |
| Bacterially expressed N-terminal fragment of recombinant human Drebrin Antibody (DBN-N-03) | |
| Primary | |
| The mouse monoclonal Drebrin Antibody (DBN-N-03) recognizes drebrin, an approximately 100-125 kDa regulator of actin cytoskeleton. | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| DBN-N-03 | |
| Western Blot, Flow Cytometry | |
| D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1 | |
| Mouse | |
| 0.1 mL | |
| 1627 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction