missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drebrin 1 Antibody (DBN-N-03), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22344PCP
This item is not returnable.
View return policy
Description
Drebrin 1 Monoclonal specifically detects Drebrin 1 in Human samples. It is validated for Flow Cytometry.
Specifications
| Drebrin 1 | |
| Monoclonal | |
| PerCP | |
| D0S117E, Developmentally-regulated brain protein, DKFZp434D064, drebrin, drebrin 1, drebrin E, drebrin E2, drebrin-1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| The mouse monoclonal Drebrin Antibody (DBN-N-03) recognizes drebrin, an approximately 100-125 kDa regulator of actin cytoskeleton. | |
| Store at 4C in the dark. | |
| IgG2b |
| Flow Cytometry | |
| DBN-N-03 | |
| Flow Cytometry | |
| DBN1 | |
| Bacterially expressed N-terminal fragment of recombinant human Drebrin Antibody (DBN-N-03) | |
| 0.1 mL | |
| Neuroscience | |
| 1627 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction