Learn More
Drebrin Mouse anti-Human, Unconjugated, Clone: 1A12H1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67589-1-IG-150UL
Description
The Drebrin antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Drebrin. This antibody recognizes human, mouse, rat, pig antigen. The Drebrin antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Drebrin | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1A12H1 | |
| Unconjugated | |
| 150 μL | |
| 1627 | |
| IgG2a |