missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DREF Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76327H
This item is not returnable.
View return policy
Description
DREF Polyclonal antibody specifically detects DREF in Human, Mouse samples. It is validated for Western Blot, ELISA
Specifications
| DREF | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 9189 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA | |
| HRP | |
| Ac-like transposable element, ALTEDNA replication-related element binding factor, hDREF, KIAA0785dREF homolog, Putative Ac-like transposable element, putative Ac-like transposon, TRAMPDREFBED-type zinc finger domain-containing protein 1, zinc finger BED domain-containing protein 1, zinc finger, BED-type containing 1 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human ZBED1. The immunogen is located within amino acids 180 - 230 of ZBED1. Amino Acid Squence: STDMWRSENQNRAYV | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction