missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ DRG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30170
This item is not returnable.
View return policy
Description
DRG1 Polyclonal antibody specifically detects DRG1 in Human samples. It is validated for Immunoprecipitation
Specifications
| DRG1 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunoprecipitation | |
| Unconjugated | |
| developmentally regulated GTP binding protein 1, developmentally regulated GTP-binding protein 1, developmentally-regulated GTP-binding protein 1, DKFZp434N1827, DRG-1, NEDD3, NEDD-3, Neural precursor cell expressed developmentally down-regulated protein 3, neural precursor cell expressed, developmentally down-regulated 3 | |
| Between 150 and 200 | |
| 100 μg | |
| Cancer, Cell Biology, Cell Cycle and Replication, GPCR, Signal Transduction | |
| 4733 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction