missing translation for 'onlineSavingsMsg'
Learn More

DRGX Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA560570

Additional Details : Weight : 0.02000kg

Product Code. 15728401

  • € 514.00 / 100µL
Estimated Shipment: 12-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: PMGLSFLPTY GCQSNRTASV ATLRMKAREH SEAVLQSANL LPSTSSSPGP VAKPAPPDGS QEKTSPTKEQ SEAEKSV Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.

DRGX Polyclonal antibody specifically detects DRGX in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

DRGX
Polyclonal
Unconjugated
DRGX
dorsal root ganglia homeobox; dorsal root ganglia homeobox protein; Dorsal root ganglion 11; drg11; Drgx; Homeobox protein DRG11; paired homeodomain protein DRG11; paired related homeobox protein-like 1; paired related homeobox protein-like 1b; paired related homeobox-like 1; paired-like homeodomain trancription factor DRG11; paired-like homeodomain transcription factor DRG11; paired-related homeobox protein-like 1; Prrxl1
Rabbit
Antigen affinity chromatography
RUO
644168
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
A6NNA5
DRGX
Recombinant protein corresponding to Human DRGX
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Research Use Only