Learn More
Schneider Gerd™ Drigalski Spatula
Product Code. 10356274
Change view
Click to view available options
Length (Metric):
165 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10356274 165 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10356274 Supplier Schneider Gerd™ Supplier No. 3532
Order Info
LengthMetric
Specifications
Quantity 1 Pk.
Product Type Drigalski Spatula
Width (Metric) Blade 30 mm
Dimensions (L x W) 165 x 30 x 2 mm
Length (Metric) 165 mm