Learn More
Invitrogen™ DRIL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596754
Description
Immunogen sequence: PYECEKRGLS NPNELQAAID SNRREGRRQS FGGSLFAYSP GGAHGMLSSP KLPVSSLGLA ASTNGSSITP APKIKKEEDS AIPITVPGRL PVSLAGHPVV AAQAAAVQAA AAQAAVAAQA AALEQLREKL ESAEPPEKKM ALVADEQQRL MQRALQQNFL AMAAQLPMSI RINSQASESR QDSAVNLTGT NGSNSISMSV EINGIMYTGV LFAQPPAPTP TSAPNKGGGG GGGSSSNAGG RGGNTGTSGG QAGPAGLSTP STSTSNNSLP; Positive Samples: Jurkat, MCF7, Mouse testis; Cellular Location: Cytoplasm, Nucleus.
The mouse Bright (B-cell regulator of IgH transcription) protein is expressed specifically in B cells and transactivates an IgH enhancer in transient transfection assays. Bright and the Drosophila 'dead ringer' (dri) gene product belong to a protein family whose members share a conserved DNA-binding domain termed the 'A/T-rich interaction domain' (ARID).
Specifications
| DRIL1 | |
| Polyclonal | |
| Unconjugated | |
| ARID3A | |
| ARID domain-containing 3A; ARID domain-containing protein 3A; ARID3A; AT rich interactive domain 3A (Bright like); AT rich interactive domain 3A (BRIGHT- like) protein; AT rich interactive domain 3A (BRIGHT-like); AT-rich interaction domain 3A; AT-rich interactive domain-containing protein 3A; B-cell regulator of IgH transcription; BRIGHT; dead ringer homolog 1; dead ringer-like 1; dead ringer-like protein 1; Dri1; DRIL1; DRIL3; DRX; E2F-binding protein 1; E2FBP1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 13496, 1820, 314616 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1.1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q62431, Q99856 | |
| ARID3A | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 324-593 of human ARID3A (NP_0052151). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.