Learn More
Kautex™ Dropper Cap & Nozzle, Narrow Neck LPDE Bottle
Product Code. 10199230
Click to view available options
For Use With (Equipment):
10/20/30 mL Bottle
1000/2000 mL Bottle
50/100/200 mL Bottle
500 mL Bottle
Diameter (Metric):
14 mm
18 mm
25 mm
28 mm
Unit Size:
Each
Pack of 100
Product Code. 10199230
Brand: Kautex™ 2000072100
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Top off your narrow neck LDPE bottles with the Kautex™ Dropper Cap & Nozzle for Narrow Neck LPDE Bottles. Closures and bottles are packed together as one unit to simplify ordering
  • Color: Natural
  • LPDE
  • Dropper includes nozzle
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Includes Nozzle
Thread Size (Metric) 00 mm
Neck Finish 00 mm
Product Type Dropper Cap
For Use With (Equipment) 500 mL Bottle
Diameter (Metric) 25 mm