Learn More
Witeg™ Dropping Funnel
Product Code. p-8010335
Click to view available options
Capacity (Metric):
25 mL
50 mL
100 mL
250 mL
500 mL
Joint Number:
14/23
29/32
Stopcock:
Yes
Yes, PTFE
Unit Size:
Each
Product Code. 10178391
Brand: Witeg™ 1 856 050
Witeg™ Dropping Funnels are fitted with a stopcock to transfer fluids in a controlled, dropwise manner.
  • Dropping funnels, acc. to DIN 12567
  • Cylindrical
  • Pressure equalizing tube
  • Graduated
  • ST-stopcock
  • Screw-thread retaining nut
  • Borosilicate glass construction
Specifications
Capacity (Metric) 50 mL
Certifications/Compliance DIN 12567
Graduated Yes
Includes Screwthreaded retaling nut
Graduations 1 mL
Joint Number 29/32
Stopcock Yes
Product Type Dropping Funnel