missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Drosha Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99092R
This item is not returnable.
View return policy
Description
Drosha Polyclonal antibody specifically detects Drosha in Human samples. It is validated for Western Blot
Specifications
| Drosha | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| drosha, double-stranded RNA-specific endoribonuclease, drosha, ribonuclease type III, EC 3.1.26.3, Etohi2, nuclear RNase III Drosha, p241, Protein Drosha, putative protein p241 which interacts with transcription factor Sp1, putative ribonuclease III, RANSE3L, ribonuclease 3, Ribonuclease III, ribonuclease III, nuclear, RN3nuclear, RNase III | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Drosha. | |
| 0.1 mL | |
| Epigenetics | |
| 29102 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction