missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Drosha Recombinant Rabbit Monoclonal Antibody (23GB5205), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553863
This item is not returnable.
View return policy
Description
Members of the ribonuclease III superfamily of double-stranded (ds) RNA-specific endoribonucleases participate in diverse RNA maturation and decay pathways in eukaryotic and prokaryotic cells (Fortin et al., 2002). The RNase III Drosha is the core nuclease that executes the initiation step of microRNA (miRNA) processing in the nucleus (Lee et al., 2003).
Specifications
| Drosha | |
| Recombinant Monoclonal | |
| Unconjugated | |
| DROSHA | |
| 1110013A17Rik; AI874853; Drosha; drosha ribonuclease III; drosha, double-stranded RNA-specific endoribonuclease; drosha, ribonuclease type III; ethanol induced 2; ETOHI2; HSA242976; MGC115770; nuclear RNase III Drosha; OTTHUMP00000220910; OTTHUMP00000220911; OTTHUMP00000220912; OTTHUMP00000220913; p241; Protein Drosha; putative protein p241 which interacts with transcription factor Sp1; putative ribonuclease III; RANSE3L; RGD1307626; ribonuclease 3; Ribonuclease III; ribonuclease III, nuclear; ribonuclease type III, nuclear; RN3; RNase III; RNASE3L; RNASEN | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 14000, 29102, 310159 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB5205 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5HZJ0, Q9NRR4 | |
| DROSHA | |
| A synthesized peptide derived from human Drosha (20-77AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction