missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DRP1 Antibody (4E11B11), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80702AF488
This item is not returnable.
View return policy
Description
DRP1 Monoclonal antibody specifically detects DRP1 in Human, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| DRP1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 4E11B11 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| DLP1, DRP1Dnm1p/Vps1p-like protein, DVLPEC 3.6.5.5, Dymple, DYMPLEDYNIV-11, dynamin 1-like, Dynamin family member proline-rich carboxyl-terminal domain less, dynamin-1-like protein, Dynamin-like protein, Dynamin-like protein 4, Dynamin-like protein IV, Dynamin-related protein 1, FLJ41912, HDYNIV, VPS1 | |
| Partial recombinant human DRP1 (between residues 100-250) expressed in E. coli. [Uniprot: O00429] | |
| 0.1 mL | |
| Apoptosis, Cell Biology, Mitochondrial Fusion Proteins, Mitochondrial Mediated Pathway, Neurodegeneration, Neuroscience, Signal Transduction | |
| 10059 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction