Learn More
Invitrogen™ DRP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570650
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 86%; Dog: 86%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 79%; Rabbit: 93%; Rat: 86%.
Members of the dystrophin family of proteins perform a critical role in the maintenance of membrane-associated complexes at points of intercellular contact in vertebrate cells. Dystrophin related protein 2 is predicted to resemble certain short C-terminal isoforms of dystrophin and dystrophin-related protein 1 (DRP1 or utrophin). DRP2 is expressed principally in the brain and spinal cord.
Specifications
| DRP2 | |
| Polyclonal | |
| Unconjugated | |
| Drp2 | |
| AW495265; DRP2; DRP-2; dystrophin related protein 2; dystrophin-related protein 2; dystrophin-related protein 2 A-form | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 1821 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q13474 | |
| Drp2 | |
| synthetic peptide directed towards the middle region of human DRP2 (aa 178-227). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |